Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ E 64d
GREENER_CHOICESDP

Catalog No. 45451 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-451 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-451 Supplier R&D Systems™ Supplier No. 4545/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cathepsin inhibitor; interferes with autolysosomal digestion

Inhibitor of cathepsins B and L; also thought to inhibit calpain. Inhibits lysosomal proteases and interferes with autolysosomal digestion when used in combination with pepstatin A (Cat. No. 1190). Lysosome and cell permeable.

Chemical Identifiers

CAS 88321-09-9
Molecular Formula C17H30N2O5

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 342.43
Purity >98%
Molecular Weight (g/mol) 342.436
Chemical Name or Material E 64d
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.