Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ E 7820
GREENER_CHOICE

Catalog No. 678510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-851-0 10 mg
67-855-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-851-0 Supplier Tocris Bioscience™ Supplier No. 6785/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

α2 integrin inhibitor; anti-angiogenic

E 7820 is a α2 integrin inhibitor. Inhibits angiogenesis in vitro (IC50 = 0.11 - 0.25 μM). Also inhibits growth of human colorectal cancer cell lines. Enhances efficacy of erlotinib in a non-small-cell lung cancer xenograft model. Also acts as a molecular glue to induce proteosomal degradation of mRNA splicing factor RBM39.

Specifications

Chemical Name or Material 3-Cyano-N-(3-cyano-4-methyl-1 H-indol-7-yl)benzenesulfonamide
CAS 289483-69-8
Quantity 10 mg
Target Integrin Receptor Inhibitors
Molecular Formula C17H12N4O2S
Formula Weight 336.37
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.