Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-Cadherin Antibody (1E1), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315180
Description
E-Cadherin Monoclonal antibody specifically detects E-Cadherin in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
E-Cadherin | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4), 150mM NaCl, and 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry | |
10 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin | |
A synthesized peptide derived from human E-Cadherin. UniProt ID (P12830). | |
100 μg | |
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction | |
999 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction