Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Cadherin Antibody (CDH1/1525), PE/Cy7, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP254551PECY7

Catalog No. N25451PECY7


Only null left
Add to Cart

Description

Description

E-Cadherin Monoclonal specifically detects E-Cadherin in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry.
Specifications

Specifications

E-Cadherin
Monoclonal
PE-Cyanine7
PBS with 0.05% Sodium Azide
CDH1
Recombinant human E-Cadherin protein (Uniprot: P12830)
0.1 mL
Primary
Human, Mouse (Negative), Rat (Negative)
IgG1 κ
Flow Cytometry
CDH1/1525
Flow Cytometry
Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin
Mouse
Protein A or G purified
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction
999.0
Store at 4°C in the dark. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.