Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-Cadherin Antibody (CDH1/2208R), Alexa Fluor™ 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP307903AF405
Description
E-Cadherin Monoclonal antibody specifically detects E-Cadherin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
E-Cadherin | |
Monoclonal | |
Alexa Fluor 405 | |
Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin | |
Recombinant full-length human E-Cadherin protein (Uniprot: P12830) | |
100 μg | |
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction | |
999 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
CDH1/2208R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction