Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Cadherin Rabbit anti-Human, Non-human Primate, Polyclonal, FabGennix

Catalog No. 502206841
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6841 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6841 Supplier Fabgennix Inc Supplier No. ECAD131AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

E-Cadherin Polyclonal antibody specifically detects E-Cadherin in Human, Non-human Primate samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen E-Cadherin
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene CDH1
Gene Accession No. P12830
Gene Alias AA960649; ARC-1; cadherin 1; cadherin 1, E-cadherin (epithelial); cadherin 1, epithelial; cadherin 1, type 1; cadherin 1, type 1, E-cadherin (epithelial); cadherin e; cadherin-1; cadherin-E; calcium-dependent adhesion protein, epithelial; CAM 120/80; CD324; CDH1; CDHE; cell adhesion molecule; cell-CAM 120/80; ECAD; E-cad; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; Ecadherin; E-cadherin; E-cadherin 1; E-Cadherin phospho; E-cadherin precursor; Epithelial cadherin; hab; half baked; LCAM; L-CAM; phospho cadherin 1; phospho E-Cadherin; phospho LCAM; phospho S838; phospho UVO; S840; S840 E-Cadherin; Ser840; Ser840 E-Cadherin; Um; UVO; Uvomorulin
Host Species Rabbit
Immunogen Multiple synthetic peptides taken within amino acid regions 50-100, 350-400, and 650-700 on human E-cadherin 1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 468009, 999
Target Species Human, Non-human Primate
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.