Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E. Coli 015 Antibody (15/443), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064541B
Description
E. Coli 015 Monoclonal antibody specifically detects E. Coli 015 in Bacteria samples. It is validated for ELISA, ImmunofluorescenceSpecifications
E. Coli 015 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Bacteria | |
Purified |
ELISA, Immunofluorescence | |
15/443 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Escherichia coli | |
Native, NCTC 9015 | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction