Learn More
Novus Biologicals E. coli Aconitate hydratase B ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Range | 78.1-5000 pg/mL |
| Assay Sensitivity | 32.4 pg/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Cell Lysates |
| Specificity | Recognizes both recombinant and natural E. coli Aconitate hydratase B. |
| For Use With (Application) | ELISA, Sandwich ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Storage is content dependent. |
| Synonym | acnB, hypothetical protein, yacI, yacJ |
| Target | E. coli |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.