Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ E. coli anti-poly-ADP-ribose binding reagent, Polyclonal,
E. coli Polyclonal Antibody
Supplier: MilliporeSigma™ MABE1031
Description
Specifically detects poly-ADP-ribose binding reagent in Human, Mouse samples, and it is validated for Dot Blot
Anti-poly-ADP-ribose binding reagent, is a His-tagged recombinant protein fused to rabbit Fc tag, expressed in and purified from Rosetta™(DE3)pLysS strain of E. coli. Anti-poly-ADP-ribose binding reagent is useful for the affinity detection of oligo- and poly-ADP-ribosylated (PARylated) proteins on membranes or on fixed cells in a manner similar to antibody-based Western blot, dot blot, and immunocytochemistry applications. The rabbit Fc tag allows visualization of the binding/labeling with conjugated anti-rabbit secondary antibodies. The Fc tag also allows Anti-poly-ADP-ribose binding reagent to be captured on Protein A resins for affinity pull-down applications.
Specifications
poly-ADP-ribose binding reagent | |
Polyclonal | |
E. coli | |
200 μL | |
Epigenetics and Nuclear Function | |
Human, Mouse | |
Purified |
Dot Blot | |
Purified from E. coli by Ni-NTA agarose. Supplied in buffer containing 10mM Tris pH 7.5, 0.2M NaCl, 10% Glycerol, 10mM Imidazole, 1mM PMSF, 1mM β-Mercaptoethanol, 10% glycerol without preservatives. | |
Ni-NTA agarose | |
RUO | |
Primary | |
Stable for one year at -80°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -80°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction