Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E. coli bepA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117604
Description
This antibody has been tested in direct-ELISA.
Functions as both a chaperone and a metalloprotease. Maintains the integrity of the outer membrane by promoting either the assembly or the elimination of outer membrane proteins, depending on their folding state. Promotes disulfide rearrangement of LptD during its biogenesis, and proteolytic degradation of LptD and BamA when their proper assembly is compromised. May facilitate membrane attachment of LoiP under unfavorable conditions.
Specifications
E. coli bepA | |
Polyclonal | |
Unconjugated | |
bepA | |
bepA | |
Recombinant Escherichia coli Beta-barrel assembly-enhancing protease protein (214-487aa). | |
100 μL | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Western Blot | |
2.625 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P66948 | |
Rabbit | |
Protein G | |
RUO | |
57732404 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction