Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E. coli Beta-lactamase CTX-M-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117577
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117577 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117577 Supplier Invitrogen™ Supplier No. PA5117577
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Broad spectrum beta-lactamase which confers resistance to penicillins, as well as first, second and third-generation cephalosporins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E. coli Beta-lactamase CTX-M-1
Applications Western Blot
Classification Polyclonal
Concentration 3.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene bla
Gene Accession No. P28585
Gene Symbols bla
Host Species Rabbit
Immunogen Recombinant Escherichia coli Beta-lactamase CTX-M-1 protein (29-291aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 39692553
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.