Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. coli CLPS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117511
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117511 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117511 Supplier Thermo Scientific Supplier No. PA5117511
Add to Cart
Add to Cart

E. coli CLPS Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen E. coli CLPS
Applications Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias 2200003J09Rik; Clps; COL; Colipase; colipase, pancreatic; COLQ; pancreatic colipase preproprotein
Host Species Rabbit
Immunogen Recombinant Escherichia coli ATP-dependent Clp protease adapter protein ClpS protein (1-105aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.