Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E. coli csrA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117654
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117654 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117654 Supplier Invitrogen™ Supplier No. PA5117654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

csrA affects glycogen biosynthesis, gluconeogenesis, cell size and surface properties. Regulates glycogen synthesis under both aerobic and anaerobic conditions. Seems to accelerate the degradation of glg gene transcripts, potentially through selective RNA binding. Acts to inhibit interaction between the LetD protein and the A subunit of DNA gyrase. Also required for motility and flagellum biosynthesis through the post-transcriptional activation of flhDC expression. This involves binding to and stabilization of the flhDC message by CsrA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E. coli csrA
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene csrA
Gene Accession No. B1XCM4
Gene Symbols csrA
Host Species Rabbit
Immunogen Recombinant Escherichia coli Translational regulator CsrA protein (1-61aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58460139
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.