Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E. coli dcyD Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117514
Description
This antibody has been tested in direct-ELISA.
Catalyzes the alpha,beta-elimination reaction of D-cysteine and of several D-cysteine derivatives. It could be a defense mechanism against D-cysteine.
Specifications
E. coli dcyD | |
Polyclonal | |
Unconjugated | |
dcyD | |
dcyD | |
Recombinant Escherichia coli D-cysteine desulfhydrase protein (2-328aa). | |
100 μg | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P76316 | |
Rabbit | |
Protein G | |
RUO | |
946831 | |
-20°C or -80°C if preferred | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction