Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E.coli DH-5 Alpha Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984447
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4447 Supplier Bioss Supplier No. BS2033R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody reacts with Escherichia coli HD5a (E.coli HD5a), Escherichia coli is a gram-negative bacillus that belongs to a larger group of Enterobacteriae - bacteria that inhabit the gastrointestinal tract. Although usually a harmless resident of the gut, some strains have the potential to cause serious problems, especially where there is an immature immune system or immunosuppression, or where the subtype of organism has acquired the ability to produce pathogenic toxins. E. coli DH5_ strain is an engineering strain widely used in biopharmaceuticals. According to the requirements of product quality inspection, it is necessary to detect the residual amount of DH5_ bacterial protein in biological products. This product is an anti-serum obtained by immunizing rabbits with DH5_ bacterial protein as an antigen, and purified by affinity chromatography to obtain antibody IgG, which can be used for ELISA, Western Blot and other experimental tests.

Specifications

Antigen E.coli DH-5 Alpha
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen Whole cell protein of Ecoli DH-5 Alpha.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.