Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E. Coli K88A Antibody (1050/66), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064537AF594
Description
E. Coli K88A Monoclonal specifically detects E. Coli K88A in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
E. Coli K88A | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
In a simple ELISA, using Biogenesis test groups, this is only reactive with E. coli K88a. Reactive with isolates fixed by phenol, formalin and boiling. | |
Store at 4°C in the dark. |
ELISA, Immunocytochemistry, Immunofluorescence | |
1050/66 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Escherichia coli | |
NCTC 10964 (Nobivac) | |
Primary | |
Bacteria | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction