Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E. Coli K88A Antibody (1050/66) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.00
Specifications
Antigen | E. Coli K88A |
---|---|
Clone | 1050/66 |
Concentration | 0.94 mg/mL |
Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
Applications | Immunocytochemistry, Immunofluorescence, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017598
![]() |
Novus Biologicals
NB100645370.025MG |
0.025 mg |
Each for $206.00
|
|
|||||
NB10064537
![]() |
Novus Biologicals
NB10064537 |
0.1 mg | N/A | N/A | N/A | ||||
Description
E. Coli K88A Monoclonal specifically detects E. Coli K88A in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
E. Coli K88A | |
0.94 mg/mL | |
Immunocytochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Mouse | |
Bacteria | |
Escherichia coli | |
NCTC 10964 (Nobivac) | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1050/66 | |
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
P04738 | |
faeC | |
IgG1 | |
Protein A purified | |
In a simple ELISA, using Biogenesis test groups, this is only reactive with E. coli K88a. Reactive with isolates fixed by phenol, formalin and boiling. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title