Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E. Coli K88A Antibody (1050/66), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064537FR
Description
E. Coli K88A Monoclonal specifically detects E. Coli K88A in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
E. Coli K88A | |
Monoclonal | |
DyLight 680 | |
Escherichia coli | |
Mouse | |
Protein A purified | |
RUO | |
In a simple ELISA, using Biogenesis test groups, this is only reactive with E. coli K88a. Reactive with isolates fixed by phenol, formalin and boiling. | |
Store at 4C in the dark. | |
IgG1 |
ELISA, Immunocytochemistry, Immunofluorescence | |
1050/66 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
faeC | |
NCTC 10964 (Nobivac) | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction