Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E. Coli Labile Toxin A Chain Antibody (20/480), DyLight 550 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064577R
Description
E. Coli Labile Toxin A Chain Monoclonal antibody specifically detects E. Coli Labile Toxin A Chain in Bacteria samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
E. Coli Labile Toxin A Chain | |
Monoclonal | |
DyLight 550 | |
Escherichia coli | |
Native, labile toxin | |
0.2 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunofluorescence | |
20/480 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction