Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E. Coli O157 Antibody (9.88), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | E. Coli O157 |
---|---|
Clone | 9.88 |
Concentration | 0.2 mg/mL |
Dilution | ELISA 1 - 5 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml |
Applications | ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25314620
![]() |
Novus Biologicals
NBP25314620UG |
20 μg |
Each for $268.00
|
|
|||||
NB253146100
![]() |
Novus Biologicals
NBP253146100UG |
100 μg |
Each for $524.00
|
|
|||||
NB253146200
![]() |
Novus Biologicals
NBP253146200UG |
200 μg | N/A | N/A | N/A | ||||
Description
E. Coli O157 Monoclonal specifically detects E. Coli O157 in E. coli samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
E. Coli O157 | |
0.2 mg/mL | |
ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
E. coli | |
A crude sonicate of E. coli O157 | |
Primary | |
Store at 4C. |
9.88 | |
ELISA 1 - 5 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml | |
Monoclonal | |
Supernatant | |
RUO | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
IgM κ | |
Tissue culture supernatant |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title