Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E. coli O157:H7 ompC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117701
Description
This antibody has been tested in direct-ELISA.
Forms passive diffusion pores which allow small molecular weight hydrophilic materials across the outer membrane.
Specifications
E. coli O157:H7 ompC | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Western Blot | |
4.555 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Recombinant Escherichia coli O157:H7 Outer membrane protein C protein (22-367aa). | |
100 μL | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction