Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E. coli RecQ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117513
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117513 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117513 Supplier Invitrogen™ Supplier No. PA5117513
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Involved in the RecF recombination pathway; its gene expression is under the regulation of the SOS system. It is a DNA helicase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E. coli RecQ
Applications Western Blot
Classification Polyclonal
Concentration 2.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene recQ
Gene Accession No. P15043
Gene Symbols recQ
Host Species Rabbit
Immunogen Recombinant Escherichia coli ATP-dependent DNA helicase RecQ protein (2-606aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58462160
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.