Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E. coli serotype O/K Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PA173031
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA173031 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA173031 Supplier Invitrogen™ Supplier No. PA173031
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73031 antibody reacts with all 'O' and 'K' antigenic serotypes of E. coli. This antibody is unabsorbed and will cross-react with related Enterobacteriaceae. PA1-73031 has been successfully used in ELISA applications. The PA1-73031 immunogen is a mixture of all antigenic serotypes of Escherichia coli. Recommended short term (< 6 months) storage is liquid at 4°C. For long term storage, aliquot and freeze.

Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E. coli serotype O/K
Applications ELISA
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Biotin
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Escherichia coli
Host Species Rabbit
Immunogen Mixture of all antigenic serotypes of Escherichia coli.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.