Learn More
Invitrogen™ E. coli serotype O Monoclonal Antibody (HEC10-189.4), Invitrogen™
Shop All Thermo Scientific Products

Description
MA1-10694 detects E. coli from E. coli O2a/2b, O7, O11, O18, O44, O75, O112, and O125 samples. MA1-10694 has been successfully used in ELISA applications. The MA1-10694 immunogen is E. coli serotypes: O18, O44, O112, O125.
Specifications
Specifications
| Antigen | E. coli serotype O |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | HEC10-189.4 |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | Escherichia coli |
| Host Species | Mouse |
| Immunogen | E. coli serotypes: O18, O44, O112, O125. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.