Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E. coli (strain K12) grxD Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117619
Description
This antibody has been tested in direct-ELISA.
Monothiol glutaredoxin involved in the biogenesis of iron-sulfur clusters.
Specifications
| E. coli (strain K12) grxD | |
| Polyclonal | |
| Unconjugated | |
| grxD | |
| Glutaredoxin 4; Grx4; Monothiol glutaredoxin | |
| Rabbit | |
| Protein G | |
| RUO | |
| 58462457, 82261796 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 2.592 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P0AC69 | |
| grxD | |
| Recombinant Escherichia coli Glutaredoxin 4 protein (1-115aa). | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction