Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E. coli (strain K12) PdeH Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144479
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144479 Supplier Invitrogen™ Supplier No. PA5144479
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

E. coli (strain K12) PdeH Polyclonal Antibody for Western Blot, ELISA

Involved in the control of the switch from cell motility to adhesion via regulation of cellular levels of cyclic-di-GMP (c-di-GMP) (PubMed:18765794). Part of a signaling cascade that regulates curli biosynthesis. The cascade is composed of two c-di-GMP control modules, in which c-di-GMP controlled by the DgcEPdeH pair (module I) regulates the activity of the DgcMPdeR pair (module II), which in turn regulates activity of the transcription factor MlrA and expression of the master biofilm regulator csgD (PubMed:23708798). Effect on flagella is controlled via the c-di-GMP-binding flagellar brake protein YcgR (PubMed:18765794).
TRUSTED_SUSTAINABILITY

Specifications

Antigen E. coli (strain K12) PdeH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene pdeH
Gene Accession No. P37646
Gene Alias Cyclic di-GMP phosphodiesterase
Gene Symbols pdeH
Host Species Rabbit
Immunogen Recombinant Escherichia coli (strain K12) YHJH protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57729221, 75201973
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.