Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E. coli (strain K12) ppsA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117576
Description
This antibody has been tested in direct-ELISA.
Catalyzes the phosphorylation of pyruvate to phosphoenolpyruvate.
Specifications
E. coli (strain K12) ppsA | |
Polyclonal | |
Unconjugated | |
ppsA | |
ppsA | |
Recombinant Escherichia coli Phosphoenolpyruvate synthase protein (2-792aa). | |
100 μL | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Western Blot | |
2.71 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P23538 | |
Rabbit | |
Protein G | |
RUO | |
946209 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction