Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E. coli tar Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117610
Description
This antibody has been tested in direct-ELISA.
Receptor for the attractant L-aspartate and related amino and dicarboxylic acids. Tar also mediates taxis to the attractant maltose via an interaction with the periplasmic maltose binding protein. Tar mediates taxis away from the repellents cobalt and nickel. Chemotactic-signal transducers respond to changes in the concentration of attractants and repellents in the environment, transduce a signal from the outside to the inside of the cell, and facilitate sensory adaptation through the variation of the level of methylation. Attractants increase the level of methylation while repellents decrease the level of methylation, the methyl groups are added by the methyltransferase CheR and removed by the methylesterase CheB.
Specifications
E. coli tar | |
Polyclonal | |
Unconjugated | |
tar | |
tar | |
Recombinant Escherichia coli Methyl-accepting chemotaxis protein II protein (212-553aa). | |
100 μg | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Western Blot | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P07017 | |
Rabbit | |
Protein G | |
RUO | |
946399 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction