Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-Selectin/CD62E Antibody (208), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289432PE
Description
E-Selectin/CD62E Monoclonal antibody specifically detects E-Selectin/CD62E in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
E-Selectin/CD62E | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
208 | |
Immunocytochemistry/ Immunofluorescence | |
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human E-Selectin/CD62E (Accession#: NP_000441.2; Met1-Pro556). | |
0.1 mL | |
Immunology, Lipid and Metabolism | |
6401 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction