Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Selectin/CD62E Antibody (40), FITC
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP289430F

Catalog No. NB066827


Description

Description

E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications

Specifications

E-Selectin/CD62E
Monoclonal
FITC
PBS
Rabbit
Protein A purified
RUO
Primary
No cross-reactivity in ELISA with: Human CD31/ PECAM1 Human VCAM1/ CD106 Human ICAM1/ CD54 Mouse VCAM1/ CD106 Mouse ICAM-1/ CD54
Store at 4°C in the dark.
IgG
ELISA
40
ELISA, Sandwich ELISA Detection
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E
This antibody was obtained from a rabbit immunized with purified, recombinant Human E-Selectin/CD62E (NP_000441.2; Met 1-Pro 556).
0.1 mL
Immunology, Lipid and Metabolism
6401
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.