Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-Selectin/CD62E Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP14554550UG
Description
E-Selectin/CD62E Polyclonal antibody specifically detects E-Selectin/CD62E in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
E-Selectin/CD62E | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2), 5 mM EDTA, 0.5% BSA, 30% Glycerol | |
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
Synthetic peptide surrounding amino acid 584 of human E-Selectin. | |
50 μg | |
Immunology, Lipid and Metabolism | |
6401 | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 0.5-4 ug/ml, Simple Western 1:50, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 10-20 ug/ml, Immunohistochemistry-Paraffin 10-20 ug/ml | |
P16581 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction