Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-Selectin/CD62E Antibody (RM0034-9J12) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $562.50
Specifications
Antigen | E-Selectin/CD62E |
---|---|
Clone | RM0034-9J12 |
Concentration | 0.5 mg/mL |
Dilution | Flow Cytometry 1:100-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:400 |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBT85473100
![]() |
Novus Biologicals
NB11085473100UG |
100 μg |
Each for $562.50
|
|
|||||
NB018041
![]() |
Novus Biologicals
NB110854730.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
Description
E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
E-Selectin/CD62E | |
0.5 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rat | |
A 0.2 um filtered solution in PBS with No Preservative | |
6401 | |
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with cytokine stimulated-mouse endothelial membrane protein. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
RM0034-9J12 | |
Flow Cytometry 1:100-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:400 | |
Monoclonal | |
Purified | |
Mouse | |
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
SELE | |
IgG2 | |
Protein G purified | |
This is specific for mouse E-Selectin |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title