Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-Selectin/CD62E Antibody (RM0034-9J12), PE/Cy5.5, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11085473PECY55
Description
E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Flow Cytometry.Specifications
E-Selectin/CD62E | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
SELE | |
Cytokine-stimulated mouse E-Selectin/CD62E | |
0.1 mL | |
Primary | |
This is specific for mouse E-Selectin | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
RM0034-9J12 | |
Flow Cytometry | |
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
Rat | |
Protein G purified | |
Immunology, Lipid and Metabolism | |
6401.0 | |
Mouse | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction