Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-Selectin/CD62E Antibody (RM0034-9J12), PE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11085473PE
Description
E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Flow Cytometry.Specifications
E-Selectin/CD62E | |
Monoclonal | |
PE | |
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
This is specific for mouse E-Selectin | |
Store at 4C in the dark. | |
IgG2 |
Flow Cytometry | |
RM0034-9J12 | |
Flow Cytometry | |
SELE | |
Cytokine-stimulated mouse E-Selectin/CD62E | |
0.1 mL | |
Immunology, Lipid and Metabolism | |
6401 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction