Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-Selectin/CD62E Rabbit anti-Mouse, Clone: 15, Novus Biologicals™

Rabbit Monoclonal Antibody
$546.00
Specifications
Antigen | E-Selectin/CD62E |
---|---|
Clone | 15 |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Western Blot.Specifications
E-Selectin/CD62E | |
Western Blot 1:500-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
Recombinant Mouse E-Selectin/CD62E Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
15 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Immunology, Lipid and Metabolism | |
0.2 um filtered solution in PBS | |
6401 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title