Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD62E/CD62P Monoclonal Antibody (1.2B6), APC
GREENER_CHOICE

Catalog No. 501137714
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-113-7714 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-113-7714 Supplier Invitrogen™ Supplier No. A15438
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD62E/CD62P Monoclonal Antibody for Flow

CD62E represents the 97 kDa type I transmembrane glycoprotein E-selectin, also known as endothelial-leukocyte adhesion molecule-1 (ELAM-1). It is expressed on cytokine-stimulated endothelium, weakly on unactivated endothelium, and is absent on leukocytes. Like other selectins, CD62E is an adhesion molecule that contributes to the initial attachment and rolling of leukocytes on endothelial surfaces, a prerequisite for leukocyte extravasation into tissues.CD62P is a 140 kDa membrane glycoprotein formerly known as platelet activation-dependent granule membrane protein (PADGEM) or GMP-140. It is expressed on platelets, megakaryocytes, and epithelium. It is thought to mediate the adhesive interactions of neutrophils and monocytes with endothelium in inflammatory responses and of activated platelets to neutrophils and monocytes in hemostasis. The monoclonal antibody 1.2B6 recognizes a common epitope shared by CD62E and CD62P.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD62E/CD62P
Applications Flow Cytometry
Classification Monoclonal
Clone 1.2B6
Concentration 0.1 mg/mL
Conjugate APC
Formulation PBS with <0.1% sodium azide; pH 7.5
Gene SELP
Gene Accession No. P16109, P16581, P98110
Gene Alias CD62; CD62 antigen-like family member E; CD62 antigen-like family member P; CD62E; CD62P; CD62P; CD62 homolog; ELAM; ELAM1; ELAM-1; endothelial adhesion molecule 1; endothelial leukocyte adhesion molecule 1; ESEL; E-selectin; GMP140; GMP-140; GMRP; Granule membrane protein 140; granule membrane protein 140kDa; granulocyte membrane protein; GRMP; LECAM2; LECAM-2; LECAM3; LECAM-3; leukocyte endothelial cell adhesion molecule 2; leukocyte-endothelial cell adhesion molecule 2; Leukocyte-endothelial cell adhesion molecule 3; PADGEM; Platelet activation dependent granule-external membrane protein; platelet alpha-granule membrane protein; PSEL; P-selectin; P-selectin precursor; RP1-117P20.2; SELE; selectin E; selectin E (endothelial adhesion molecule 1); selectin P; selectin P (granule membrane protein 140kDa, antigen CD62); SELP
Gene Symbols SELE, SELP
Host Species Mouse
Immunogen TNF stimulated human endothelial cells.
Purification Method purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397150, 397508, 6401, 6403
Target Species Human, Pig
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.