Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-selectin Recombinant Rabbit Monoclonal Antibody (102), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
$293.50 - $565.00
Specifications
Antigen | E-selectin |
---|---|
Clone | 102 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543356
|
Invitrogen™
MA543356 |
50 μL |
Each for $293.50
|
|
|||||
PIMA543138
![]() |
Invitrogen™
MA543138 |
100 μL |
Each for $565.00
|
|
|||||
Description
Specificity: Rat E-Selectin/CD62e/SELE
Leukocytes adhere to the blood vessel endothelium during extravasation in postcapillary venules of lymph nodes. In addition, leukocyte adhesion occurs in the capillaries and small venules at any site in the body after onset of inflammation. This response is immediate and involves a cascade of adhesion receptors. At the endothelial surface members of the selectin and immunoglobulin superfamilies participate in this cascade. Selectins are C-type cell surface lectins that play a role in leukocyte adhesion to the blood vessel wall endothelium. E-selectin (CD62E) is an endothelial cell specific selectin that is expressed only after activation with proinflammatory cytokines. In vitro experiments have shown that IL-1, TNFalpha and bacterial wall components like lipopolysaccharides induce the transcription of E-selectin in a NFkB dependent signaling cascade. E-selectin has been associated with blood vessel endothelium in diverse inflammatory situations.Specifications
E-selectin | |
1 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
CD62 antigen-like family member E; CD62E; e selectin; Elam; ELAM1; ELAM-1; endothelial adhesion molecule 1; endothelial leukocyte adhesion molecule; endothelial leukocyte adhesion molecule 1; endothelial leukocyte adhesion molecule I; ESEL; E-selectin; LECAM2; leukocyte endothelial cell adhesion molecule 2; leukocyte-endothelial cell adhesion molecule 2; RP1-117P20.2; SELE; selectin E; selectin E (endothelial adhesion molecule 1); selectin, endothelial cell | |
SELE | |
IgG | |
Protein A | |
Antibody |
102 | |
ELISA | |
Unconjugated | |
Rabbit | |
Rat | |
P98105 | |
25544 | |
Recombinant Rat E-Selectin/CD62e/SELE Protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
SELE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title