Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Syt1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501564938
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4938 Supplier Bethyl Laboratories, Inc Supplier No. A303362A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

May play a role as calcium-regulated intrinsic membrane protein.

Specifications

Antigen E-Syt1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ESYT1
Gene Accession No. Q9BSJ8
Gene Alias FAM62A; family with sequence similarity 62 (C2 domain containing), member A; KIAA0747; MBC2; membrane bound C2 domain containing protein; vp115
Gene Symbols ESYT1
Host Species Rabbit
Immunogen Between 1054 and 1104
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23344
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.