Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-Tag Epitope Tag Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600527AF750
Description
E-Tag Epitope Tag Polyclonal antibody specifically detects E-Tag Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
E-Tag Epitope Tag | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
E epitope tag, GAPVPYPDPLEPR epitope tag, GAPVPYPDPLEPR tag | |
Rabbits were immunized with E-Tag (GAPVPYPDPLEPR) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. | |
0.1 mL | |
Primary | |
Purified | |
Epitope Tag |
Western Blot, ELISA, Immunofluorescence | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction