Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (1S,2R)-(+)-Ephedrine Hydrochloride, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco E0111ML

Catalog No. 01-001-277


Only null left
Add to Cart

Description

Description

Ephedrine is a methamphetamine analog commonly used as a stimulant and appetite suppressant. It is also found as an active constituent in herbs used for traditional Chinese medicine, such as má huáng. This certified reference standard can be used Snap-N-Spike™ as a starting material in the preparation of calibrators or controls for clinical or forensic applications or Snap-N-Shoot™ in quality control testing of dietary supplements containing ephedrine.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 24221-86-1
C6H5CH[CH(NHCH3)CH3]OH · HCl
UN 1230 - class 3 - PG 2 - Methanol, solution
(1S,2R)-(+)-alpha-(1-Methylaminoethyl)benzyl alcohol hydrochloride, (1S,2R)-(+)-2-Methylamino-1-phenyl-1-propanol hydrochloride, D-alpha-(1-Methylaminoethyl)benzyl alcohol hydrochloride
Ampule
9.7°C (49.5°F) - closed cup
218 to 220°C (lit.)
MFCD00064551
1.0 mg/mL in methanol (as free base)
201.69
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation