Learn More
Thermo Scientific™ E1-ClipTip™ Electronic Adjustable Tip Spacing Multichannel Equalizer Pipettes: PROMO
Catalog No. 14387965TI
Change view
Click to view available options
Volume (Metric):
1 to 30 μL
1.5 to 12.5 μL
10 to 300 μL
15 to 1250 μL
Number of Channels:
6
8
12
Compatible Tips:
ClipTip 1250
ClipTip 300
ClipTip 384 12.5
ClipTip 384 30
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels Compatible Tips
14-387-965TI 15 to 1250 μL 6 ClipTip 1250
14-387-969TI 1.5 to 12.5 μL 8 ClipTip 384 12.5
14-387-970TI 1 to 30 μL 8 ClipTip 384 30
14-387-967TI 10 to 300 μL 8 ClipTip 300
14-387-968TI 15 to 1250 μL 8 ClipTip 1250
14-387-972TI 1.5 to 12.5 μL 12 ClipTip 384 12.5
14-387-973TI 1 to 30 μL 12 ClipTip 384 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 14-387-965TI Supplier Thermo Scientific™ Supplier No. P4672090
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Innovative ClipTip interlocking tip interface technology
  • Tip is locked and sealed in place until ejected— no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
  • Tips are attached and ejected with a light touch— eliminates the frustration of banging tips on the pipette
An ideal choice for sample transfers
Reduce repetition and save time between labware formats with adjustable tip spacing in Equalizer models
Unified operation for multiple users and environments
  • Personalized user interface and password protection ensure consistent operation among multiple users.
  • Electronic operation to help you enjoy pipetting again
  • Designed for optimum comfort and ease of use, the electronic tip ejection and index finger pipetting action let your thumb relax during pipetting.
Two operating options ensure optimal function for every application
Choose a step based Matrix or Presets with settings for your most common applications
MyPipette™ - One click access for more efficient and unified pipetting
Customize your main menu view by organizing icons in your preferred order or creating shortcuts for your favorite programs and pipetting functions. You can also protect important settings from unintentional editing with a password.
Store up to 20 programs
Tired of continuous editing? The Programs function allows you to store 20 your most common Matrix or Presets protocols with a specific name.
A pipette for every application
Choose from single channels, 8-16 channel multichannels, or 6- 12-channel adjustable tip spacing Equalizer multichannel pipettes within the volume range of 0.5-1250μL.
Track your pipette
Service and calibration trackers remind you when the pipette needs to be sent to serviced
Liquid specific calibrations
Higher quality results with liquid specific calibrations
Li-Ion battery
Long lasting performance and fast recharging.
Flexible re-charging
Use either charging stand or a plug, you decide
2-year warranty
With convenient web registration
Compatible with Thermo Scientific ClipTip pipette tips.
† E1-ClipTip 12.5 and all 384 E1-ClipTip pipettes utilize snap and seal technology
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips ClipTip 1250
Description E1-ClipTip Equalizer
Adjustable Tip Spacing Yes
Color Turquoise
Model E1-ClipTip™
Number of Channels 6
Product Line E1-ClipTip
Programmability Yes
Type Electronic Pipette
Volume (Metric) 15 to 1250 μL
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.