Learn More
E2-25K Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304125A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
The protein encoded by this gene belongs to the ubiquitin-conjugating enzyme family. This protein interacts with RING finger proteins, and it can ubiquitinate huntingtin, the gene product for Huntington′s disease. Known functions for this protein include a role in aggregate formation of expanded polyglutamine proteins and the suppression of apoptosis in polyglutamine diseases, a role in the dislocation of newly synthesized MHC class I heavy chains from the endoplasmic reticulum, and involvement in foam cell formation. Multiple transcript variants encoding different isoforms have been identified for this gene.Specifications
E2-25K | |
Polyclonal | |
Unconjugated | |
UBE2K | |
UBE2K; HIP2; huntingtin interacting protein 2; LIG; HYPG; ubiquitin-protein ligase; ubiquitin carrier protein; ubiquitin-conjugating enzyme E2-25 KDA; ubiquitin-conjugating enzyme E2K (UBC1 homolog, yeast); DKFZp564C1216; DKFZp686J24237; E2-25K; | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3093, 53323 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P61086, P61087 | |
Ube2k | |
Between 150 and 200 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.