Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2-Crimson Mouse anti-Tag, HRP, Clone: OTI9B3, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA183017

Description
E2-Crimson fluorescent protein.Specifications
E2-Crimson | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide | |
Mouse | |
Affinity Chromatography | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Western Blot | |
OTI9B3 | |
HRP | |
E2-Crimson | |
Recombit protein and recognizes the E2-Crimson protein. | |
100 μL | |
Primary | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction