Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E2 tag Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116401
Description
E2 tag Polyclonal Antibody for Western Blot
The E2 Tag is a peptide which is derived from the Bovine Pappillomavirus type-1 transactivator protein E2 and consists of 10 amino acids(SSTSSDFRDR). The detection of E2-tagged proteins is based on the binding of mouse monoclonal specific to the tagged recombinant protein.
Specifications
| E2 tag | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| Tag | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| KLH conjugated synthetic peptide of E2 tag. | |
| 200 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction