Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F-1 Antibody (16G7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB600210
Description
E2F-1 Monoclonal specifically detects E2F-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
E2F-1 | |
Monoclonal | |
1mg/mL | |
Western Blot 1:500 - 1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100 - 1:1000 | |
E2F1 | |
The region encoding the DNA binding domain (amino acids 244-264) of E2F-1 expressed in E. coli. | |
100 μL | |
Apoptosis, Cancer, Cell Cycle and Replication, Transcription Factors and Regulators | |
1869 | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
16G7 | |
Unconjugated | |
E2F transcription factor 1, E2F-1, PBR3, PRB-binding protein E2F-1, RBAP1, RBAP-1, RBBP3, RBBP-3, RBP3, Retinoblastoma-associated protein 1, Retinoblastoma-binding protein 3, transcription factor E2F1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This is specific for human E2F-1. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction