Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F-1 Antibody (1D12), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | E2F-1 |
---|---|
Clone | 1D12 |
Dilution | Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
E2F-1 Monoclonal antibody specifically detects E2F-1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
E2F-1 | |
Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
E2F transcription factor 1, E2F-1, PBR3, PRB-binding protein E2F-1, RBAP1, RBAP-1, RBBP3, RBBP-3, RBP3, Retinoblastoma-associated protein 1, Retinoblastoma-binding protein 3, transcription factor E2F1 | |
A synthesized peptide derived from Human E2F-1 [UniProt Q01094] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
1D12 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1869 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title