Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F-2 Rabbit anti-Human, Mouse, Clone: 1H7D6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
| Antigen | E2F-2 |
|---|---|
| Clone | 1H7D6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
E2F-2 Monoclonal antibody specifically detects E2F-2 in Human, Mouse samples. It is validated for Western BlotSpecifications
| E2F-2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Transcription Factors and Regulators | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1870 | |
| IgG | |
| Affinity purified |
| 1H7D6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| E2F transcription factor 2, E2F-2transcription factor E2F2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 303-395 of human E2F-2 (Q14209). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title