Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F-4 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP121374C
Description
E2F-4 Polyclonal antibody specifically detects E2F-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
E2F-4 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
E2F transcription factor 4, p107/p130-binding, E2F-4p107/p130-binding protein, transcription factor E2F4 | |
The immunogen recognized by this antibody maps to a region between residue 363 and 413 of human E2F transcription factor 4 using the numbering given in entry NP_001941.2 (GeneID 1874). | |
0.1 mL | |
Cell Cycle and Replication | |
1874 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction