Learn More
E2F1 Mouse anti-Human, Unconjugated, Clone: 5D7G8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | E2F1 |
---|---|
Clone | 5D7G8 |
Concentration | 1000 μg/mL |
Applications | ChIP Assay, ELISA, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-9013
![]() ![]() |
Proteintech Group Inc
665151IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The E2F1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human E2F1. This antibody recognizes human, mouse, rat antigen. The E2F1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
E2F1 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
1869 | |
Primary |
5D7G8 | |
ChIP Assay, ELISA, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Human, Rat, Mouse | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.