Learn More
Invitrogen™ E2F1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120480
Description
Positive test controls include: mouse brain, rat brain, rat testis. The target is usually found in the following locations: Nucleus. Immunogen sequence: ETVGGISPGK TPSQEVTSEE ENRATDSATI VSPPPSSPPS SLTTDPSQSL LSLEQEPLLS RMGSLRAPVD EDRLSPLVAA DSLLEHVRED FSGLLPEEFI SLSPPHEALD YHFGLEEGEG IRDLFDCDFG DLTPLDF.
The protein encoded by this gene is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein and another 2 members, E2F2 and E2F3, have an additional cyclin binding domain. This protein binds preferentially to retinoblastoma protein pRB in a cell-cycle dependent manner. It can mediate both cell proliferation and p53-dependent/independent apoptosis.
Specifications
| E2F1 | |
| Polyclonal | |
| Unconjugated | |
| E2F1 | |
| E2F transcription factor 1; E2F1; E2F-1; mKIAA4009; PBR3; PRB-binding protein E2F-1; RBAP1; RBAP-1; RBBP3; RBBP-3; RBP3; Retinoblastoma-associated protein 1; retinoblastoma-binding protein 3; Transcription factor E2F1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13555, 1869, 399489 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.84 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O09139, Q01094, Q61501 | |
| E2F1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 301-437 of human E2F1 (NP_0052161). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.